Learn More
Medchemexpress LLC HY-107425 1mg Medchemexpress, MZ 1 CAS:1797406-69-9 Purity:>98%
SDP
Catalog No. 501968411
Encompass_Preferred
Medchemexpress, HY-107425 1mg MZ 1 CAS:1797406-69-9 MZ 1 is a BRD4 protein degrader based on PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8411 Supplier Medchemexpress LLC Supplier No. HY1074251MG
Medchemexpress, HY-107425 1mg MZ 1 CAS:1797406-69-9 MZ 1 is a BRD4 protein degrader based on PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.