Learn More
Selleck Chemical LLC MZ-1 2mg
SDP
Supplier:  Selleck Chemical LLC S88892MG
Encompass_Preferred
MZ-1 is a PROTAC degrader of bromodomain-containing protein 4 (BRD4). MZ-1 binds to the Brd bromodomain with Kd of 62 nM, 60 nM, 21 nM, 13 nM, 39 nM and 15 nM for Brd2BD1, Brd2BD2, Brd3BD1, Brd3BD2, Brd4BD1 and Brd4BD2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3419