Learn More
TARGETMOL CHEMICALS INC MZ 1 5MG
SDP
Catalog No. 502579764
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. MZ 1 is a BRD4 protein degrader based on PROTAC technology. purity: 99%
Catalog No. 50-257-9764 Supplier TARGETMOL CHEMICALS INC Supplier No. T54365MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. MZ 1 is a BRD4 protein degrader based on PROTAC technology. purity: 99%