Learn More
Selleck Chemical LLC MZ-101-E1913-25MG
SDP
Catalog No. 503135828
Encompass_Preferred
MZ-101 (GYS1-IN-2) is a potent small molecule inhibitor of Glycogen synthase 1(GYS1) with an IC50 value of 0 041 M By depleting muscle glycogen MZ-101 treatment normalizes glycogen levels and corrects transcriptional and metabolic abnormalities in Pompe mice
Catalog No. 50-313-5828 Supplier Selleck Chemical LLC Supplier No. E191325MG
MZ-101 (GYS1-IN-2) is a potent small molecule inhibitor of Glycogen synthase 1(GYS1) with an IC50 value of 0 041 M By depleting muscle glycogen MZ-101 treatment normalizes glycogen levels and corrects transcriptional and metabolic abnormalities in Pompe mice