Learn More
Electron Microscopy Sciences EMS Tweezers Style 7, 134 mm
SDP
Catalog No. 5023942
Encompass_Preferred
These tweezers have Zirconia ceramic precision tips. The surface resistively is 10E12 ohm. Contamination free, high temperature (1000 degree C). Aluminum handles. Lightweight.
OAL: 134 mm
Tips: Ceramic tip length 43.5 mm, tip width 0.6 mm
Catalog No. 50-239-42 Supplier Electron Microscopy Sciences Supplier No. 781277
These tweezers have Zirconia ceramic precision tips. The surface resistively is 10E12 ohm. Contamination free, high temperature (1000 degree C). Aluminum handles. Lightweight.
OAL: 134 mm
Tips: Ceramic tip length 43.5 mm, tip width 0.6 mm