Learn More
Electron Microscopy Sciences Style 71 MZ Ceramic Tip Tweezer (Tweezers only)
SDP
Catalog No. 5023945
Encompass_Preferred
Ceramic replaceable tip tweezers with thin, fine tips. Stainless steel is anti-acid and anti-magnetic. OAL 135 mm. Tweezers only, Tips sold separately.
Catalog No. 50-239-45 Supplier Electron Microscopy Sciences Supplier No. 7812771
Ceramic replaceable tip tweezers with thin, fine tips. Stainless steel is anti-acid and anti-magnetic. OAL 135 mm. Tweezers only, Tips sold separately.