Learn More
Electron Microscopy Sciences EMS Tweezers Style 73, 134 mm
SDP
Catalog No. 5023940
Encompass_Preferred
These tweezers have Zirconia ceramic precision tips. The surface resistively is 10E12 ohm. Contamination free, high temperature (1000 degree C). Aluminum handles. Lightweight.
OAL: 134 mm
Tips: Ceramic tip length 33 mm, tip width 0.3 mm
Catalog No. 50-239-40 Supplier Electron Microscopy Sciences Supplier No. 7812773
These tweezers have Zirconia ceramic precision tips. The surface resistively is 10E12 ohm. Contamination free, high temperature (1000 degree C). Aluminum handles. Lightweight.
OAL: 134 mm
Tips: Ceramic tip length 33 mm, tip width 0.3 mm