Learn More
DSI LLC MZ- Minizone Refiner
Catalog No. 501870024
Encompass_Preferred
MZ- Minizone Refiner; ZONE REFINING is the method of choice for ultra-purification of organic chemicals. This technique: • Requires no solvent or column
• Sample is 100% recovered
• Operates automatically
• Sample sizes from 250 mg to 4 grams are easily purified
• Requires only minutes to set up
Chemicals that melt without decomposition and are at least partially crystalline are candidates for zone refining. Applications include: • Preparation of standards
• Isolation of Impurities
• Purification of laser dyes, cintillation materials, organic semiconductors
Catalog No. 50-187-0024 Supplier DSI LLC Supplier No. MZ
Add to Cart
Add to Cart
Request a Quote
MZ- Minizone Refiner; ZONE REFINING is the method of choice for ultra-purification of organic chemicals. This technique: • Requires no solvent or column
• Sample is 100% recovered
• Operates automatically
• Sample sizes from 250 mg to 4 grams are easily purified
• Requires only minutes to set up
Chemicals that melt without decomposition and are at least partially crystalline are candidates for zone refining. Applications include: • Preparation of standards
• Isolation of Impurities
• Purification of laser dyes, cintillation materials, organic semiconductors