Learn More
Cayman Chemical MZ1, 1797406-69-9, 1 mg
SDP
Supplier:  Cayman Chemical 216221
Encompass_Preferred
A PROTAC that drives the selective proteasomal degradation of BRD4; also causes the removal of BRD2 and BRD3 when used at higher concentrations
Catalog No. NC1579723
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.