Learn More
Medchemexpress LLC HY-112377 10mg Medchemexpress, MZP-55 CAS:2010159-48-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11237710MG
Encompass_Preferred
Medchemexpress, HY-112377 10mg MZP-55 CAS:2010159-48-3 MZP-55 is a selective degrader of BRD3/4 based on PROTAC technology, with a Kd of 8 nM for Brd4BD2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9087