Learn More
Medchemexpress LLC N-(1-(1H-indol-3-yl)hexan-2-yl)-2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carboxamide (racemic) | 1802518-63-3 | 99.9% | 425.59 g·mol⁻¹ | C23H31N5OS | 25MG
SDP
Catalog No. 5000237756
Encompass_Preferred
(Rac)-Minzasolmin is a racemic small-molecule inhibitor of alpha-synuclein misfolding that penetrates the blood-brain barrier. It acts on early-stage aggregation by displacing membrane-bound oligomers, promoting reversion to monomers and reducing pathological aggregation. It is used in preclinical research to evaluate effects on alpha-synuclein deposition and neuro-pathological markers in models of Parkinson's disease and dementia with Lewy bodies.
  • Racemic small-molecule inhibitor of alpha-synuclein misfolding.
  • Blood-brain barrier penetrant for central nervous system studies.
  • Modulates early-stage alpha-synuclein aggregation mechanisms.
  • Shown to reduce alpha-synuclein deposition in preclinical models.
  • Supplied as a solid with high chemical purity for laboratory use.
  • Available in small-scale quantities suitable for preclinical experiments.
Catalog No. 50-002-37756 Supplier Medchemexpress LLC Supplier No. HY12528725MG
(Rac)-Minzasolmin is a racemic small-molecule inhibitor of alpha-synuclein misfolding that penetrates the blood-brain barrier. It acts on early-stage aggregation by displacing membrane-bound oligomers, promoting reversion to monomers and reducing pathological aggregation. It is used in preclinical research to evaluate effects on alpha-synuclein deposition and neuro-pathological markers in models of Parkinson's disease and dementia with Lewy bodies.
  • Racemic small-molecule inhibitor of alpha-synuclein misfolding.
  • Blood-brain barrier penetrant for central nervous system studies.
  • Modulates early-stage alpha-synuclein aggregation mechanisms.
  • Shown to reduce alpha-synuclein deposition in preclinical models.
  • Supplied as a solid with high chemical purity for laboratory use.
  • Available in small-scale quantities suitable for preclinical experiments.