Learn More
Medchemexpress LLC N-[1-(3,4-dimethoxyphenyl)ethylideneamino]-3-(4-methoxyphenyl)-1H-pyrazole-5-carboxamide | 1259484-97-3 | 99.3% | 394.42 g/mol | C21H22N4O4 | 5 MG
SDP
Catalog No. 5000262954
Encompass_Preferred
SKI-178 is a small-molecule inhibitor of sphingosine kinase 1 (SphK1) and sphingosine kinase 2 (SphK2) that induces apoptosis in various cancer cell lines. It is provided as a high-purity research reagent for in vitro studies, with reported cytotoxic activity in the low micromolar range.
  • Potent SphK1 and SphK2 inhibition
  • Cytotoxic in cancer cell lines at low micromolar concentrations
  • High purity suitable for research applications
  • Available in multiple package sizes for screening and follow-up studies
  • Molecular weight 394.42 and formula C21H22N4O4
Catalog No. 50-002-62954 Supplier Medchemexpress LLC Supplier No. HY128925MG
SKI-178 is a small-molecule inhibitor of sphingosine kinase 1 (SphK1) and sphingosine kinase 2 (SphK2) that induces apoptosis in various cancer cell lines. It is provided as a high-purity research reagent for in vitro studies, with reported cytotoxic activity in the low micromolar range.
  • Potent SphK1 and SphK2 inhibition
  • Cytotoxic in cancer cell lines at low micromolar concentrations
  • High purity suitable for research applications
  • Available in multiple package sizes for screening and follow-up studies
  • Molecular weight 394.42 and formula C21H22N4O4