Learn More
Medchemexpress LLC N-(1-((3-(4-(dimethylamino)butanamido)phenyl)sulfonyl)piperidin-4-yl)-4-(2,4,6-trichlorobenzamido)-1 | 2741262-15-5 | 98.8% | 685.02 g/mol | C28H32Cl3N7O5S | 10 MG
SDP
Catalog No. 5000288899
Encompass_Preferred
Small-molecule inhibitor of cyclin-dependent kinases CDK14 and CDK16 for biochemical and cell-based research. It demonstrates potent inhibition with reported IC50 values of 5.9 nM (CDK14) and 139.1 nM (CDK16). The compound is supplied as a purified solid suitable for laboratory use.
  • Potent inhibition of CDK14 and CDK16 with reported IC50s of 5.9 nM and 139.1 nM.
  • High purity (98.8% by HPLC).
  • Well-characterized physicochemical properties, molecular weight 685.02 g/mol.
  • Supplied as a solid suitable for low-temperature storage.
  • Available in milligram quantities for research use (10 MG).
Catalog No. 50-002-88899 Supplier Medchemexpress LLC Supplier No. HY16317010MG
Small-molecule inhibitor of cyclin-dependent kinases CDK14 and CDK16 for biochemical and cell-based research. It demonstrates potent inhibition with reported IC50 values of 5.9 nM (CDK14) and 139.1 nM (CDK16). The compound is supplied as a purified solid suitable for laboratory use.
  • Potent inhibition of CDK14 and CDK16 with reported IC50s of 5.9 nM and 139.1 nM.
  • High purity (98.8% by HPLC).
  • Well-characterized physicochemical properties, molecular weight 685.02 g/mol.
  • Supplied as a solid suitable for low-temperature storage.
  • Available in milligram quantities for research use (10 MG).