Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(1,3-benzothiazol-2-yl-1-oxo-propan-2-yl)-4-methoxyindole-2-carboxamide | 1471484-62-4 | MFCD34567722 | 98.8% | 575.68 g/mol | C30H33N5O5S | 5 MG
SDP

Catalog No. 5000221023
Encompass_Preferred

YH-53 is a peptidomimetic small-molecule inhibitor of 3CLpro (the SARS-CoV main protease) used for in vitro and in vivo research. It is supplied as a light yellow to green-yellow solid with characterized purity, molecular properties, solubility profiles, and recommended storage conditions for research formulation and biological evaluation.

  • Peptidomimetic 3CLpro inhibitor for SARS-CoV protease research.
  • High purity (98.8%) suitable for biochemical assays.
  • Molecular weight 575.68 g/mol; formula C30H33N5O5S.
  • Soluble in DMSO at 50 mg/mL; reported in vivo formulations ≥2.08 mg/mL.
  • Powder stability: -20°C (3 years) and 4°C (2 years); in solvent stability: -80°C (6 months).
  • Available in small research packs and as ready-to-use DMSO solutions.

Catalog No. 50-002-21023 Supplier Medchemexpress LLC Supplier No. HY1393115MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YH-53 is a peptidomimetic small-molecule inhibitor of 3CLpro (the SARS-CoV main protease) used for in vitro and in vivo research. It is supplied as a light yellow to green-yellow solid with characterized purity, molecular properties, solubility profiles, and recommended storage conditions for research formulation and biological evaluation.

  • Peptidomimetic 3CLpro inhibitor for SARS-CoV protease research.
  • High purity (98.8%) suitable for biochemical assays.
  • Molecular weight 575.68 g/mol; formula C30H33N5O5S.
  • Soluble in DMSO at 50 mg/mL; reported in vivo formulations ≥2.08 mg/mL.
  • Powder stability: -20°C (3 years) and 4°C (2 years); in solvent stability: -80°C (6 months).
  • Available in small research packs and as ready-to-use DMSO solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.