Learn More
Medchemexpress LLC N-(1,3-dimethyl-2-oxo-6-piperidin-1-ylbenzimidazol-5-yl)-2-methoxybenzamide | 901245-65-6 | MFCD14744183 | 98.3% | 394.47 g·mol-1 | C22H26N4O3 | 10 MG
SDP
Catalog No. 5000294701
Encompass_Preferred
GSK-5959 is a potent, selective, cell-permeable small-molecule inhibitor of the BRPF1 bromodomain with an IC50 of approximately 80 nM. It is used as a research tool to probe BRPF1-dependent chromatin regulation in biochemical and cellular assays. Molecular formula C22H26N4O3, molecular weight 394.47 g·mol-1, CAS 901245-65-6.
  • Potent and selective BRPF1 bromodomain inhibition.
  • Cell-permeable for cellular assay applications.
  • Suitable for biochemical assays and target validation studies.
  • Documented physicochemical identifiers for reproducibility.
  • Provided in milligram-scale quantities for research use.
Catalog No. 50-002-94701 Supplier Medchemexpress LLC Supplier No. HY1866510MG
GSK-5959 is a potent, selective, cell-permeable small-molecule inhibitor of the BRPF1 bromodomain with an IC50 of approximately 80 nM. It is used as a research tool to probe BRPF1-dependent chromatin regulation in biochemical and cellular assays. Molecular formula C22H26N4O3, molecular weight 394.47 g·mol-1, CAS 901245-65-6.
  • Potent and selective BRPF1 bromodomain inhibition.
  • Cell-permeable for cellular assay applications.
  • Suitable for biochemical assays and target validation studies.
  • Documented physicochemical identifiers for reproducibility.
  • Provided in milligram-scale quantities for research use.