Learn More
Medchemexpress LLC N-[[1-[3-(dimethylamino)propyl]indol-3-yl]methyl]decan-1-amine | 1416313-72-8 | 99.1% | C24H41N3 | 10MG
SDP
Catalog No. 5000260074
Encompass_Preferred
Dynole 2-24 is an indole-based dynamin GTPase inhibitor used in cell biology research to block dynamin-mediated endocytosis. It exhibits activity against dynamin I and II, is supplied as a high-purity research-grade small molecule, and is soluble in common organic solvents for in vitro assays.
  • Potent dynamin GTPase inhibition for cellular studies.
  • High purity appropriate for research use.
  • Oil form, colorless to light yellow, soluble in DMSO.
  • Provided in small quantities suitable for screening and mechanistic work.
  • Applicable to studies of endocytosis and membrane trafficking.
Catalog No. 50-002-60074 Supplier Medchemexpress LLC Supplier No. HY14508010MG
Dynole 2-24 is an indole-based dynamin GTPase inhibitor used in cell biology research to block dynamin-mediated endocytosis. It exhibits activity against dynamin I and II, is supplied as a high-purity research-grade small molecule, and is soluble in common organic solvents for in vitro assays.
  • Potent dynamin GTPase inhibition for cellular studies.
  • High purity appropriate for research use.
  • Oil form, colorless to light yellow, soluble in DMSO.
  • Provided in small quantities suitable for screening and mechanistic work.
  • Applicable to studies of endocytosis and membrane trafficking.