Learn More
Medchemexpress LLC N-(1,3-diphenyl-1H-pyrazol-5-yl)-4-nitrobenzamide | 890764-36-0 | MFCD12546140 | 99.6% | 384.39 g/mol | C22H16N4O3 | 25 MG
SDP
Catalog No. 5000241327
Encompass_Preferred
VU-29 (N-(1,3-diphenyl-1H-pyrazol-5-yl)-4-nitrobenzamide, CAS 890764-36-0) is a research small molecule that acts as a positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5). Supplied as a high-purity solid, it is intended for use in in vitro and in vivo pharmacology and neuroscience research.
  • Acts as a positive allosteric modulator of mGlu5 receptor.
  • High purity suitable for research applications.
  • Solid form for convenient handling and storage.
  • Available in small mass packages for preclinical studies.
  • Characterized by standard chemical identifiers for traceability.
Catalog No. 50-002-41327 Supplier Medchemexpress LLC Supplier No. HY10750825MG
VU-29 (N-(1,3-diphenyl-1H-pyrazol-5-yl)-4-nitrobenzamide, CAS 890764-36-0) is a research small molecule that acts as a positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5). Supplied as a high-purity solid, it is intended for use in in vitro and in vivo pharmacology and neuroscience research.
  • Acts as a positive allosteric modulator of mGlu5 receptor.
  • High purity suitable for research applications.
  • Solid form for convenient handling and storage.
  • Available in small mass packages for preclinical studies.
  • Characterized by standard chemical identifiers for traceability.