Learn More
Medchemexpress LLC N-(1,3-diphenyl-1H-pyrazolo-5-yl)-4-nitrobenzamide | 890764-36-0 | MFCD12546140 | 99.6% | 384.39 g·mol⁻¹ | C22H16N4O3 | 50 MG
SDP
Catalog No. 5000240578
Encompass_Preferred
VU-29 is a research-grade small molecule that functions as a positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5). It is provided as a purified solid for preclinical and in vitro pharmacology studies and assay development.
  • Acts as a positive allosteric modulator of mGlu5 receptor.
  • Reported EC50 ≈ 9 nM for rat mGlu5 and Ki ≈ 244 nM for the MPEP allosteric site.
  • High purity (~99.6%) suitable for analytical and pharmacological assays.
  • Molecular formula C22H16N4O3 and molecular weight 384.39 g·mol⁻¹.
  • Intended for research use only, not for human or veterinary applications.
  • Available in small-scale pack sizes for assay development and screening.
Catalog No. 50-002-40578 Supplier Medchemexpress LLC Supplier No. HY10750850MG
VU-29 is a research-grade small molecule that functions as a positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5). It is provided as a purified solid for preclinical and in vitro pharmacology studies and assay development.
  • Acts as a positive allosteric modulator of mGlu5 receptor.
  • Reported EC50 ≈ 9 nM for rat mGlu5 and Ki ≈ 244 nM for the MPEP allosteric site.
  • High purity (~99.6%) suitable for analytical and pharmacological assays.
  • Molecular formula C22H16N4O3 and molecular weight 384.39 g·mol⁻¹.
  • Intended for research use only, not for human or veterinary applications.
  • Available in small-scale pack sizes for assay development and screening.