Learn More
Medchemexpress LLC N-(1,3-diphenylpyrazol-5-yl)-4-nitrobenzamide | 890764-36-0 | MFCD12546140 | 99.6% | 384.39 g/mol | C22H16N4O3 | 10 MG
SDP
Catalog No. 5000240890
Encompass_Preferred
VU-29 is a research-stage positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5), used in preclinical pharmacology to selectively potentiate mGlu5 signaling. It is supplied as a powder with high chemical purity and is intended for in vitro and in vivo formulation following recommended solvent systems and storage conditions.
  • Positive allosteric modulator of mGlu5 receptor, suitable for receptor pharmacology studies.
  • High purity (99.6%) for reliable experimental results.
  • Powder form for flexible dosing and formulation.
  • Soluble in DMSO at 50 mg/mL; recommended vehicle progression for in vivo administration.
  • Stable under recommended storage conditions for extended shelf life in powder form.
Catalog No. 50-002-40890 Supplier Medchemexpress LLC Supplier No. HY10750810MG
VU-29 is a research-stage positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5), used in preclinical pharmacology to selectively potentiate mGlu5 signaling. It is supplied as a powder with high chemical purity and is intended for in vitro and in vivo formulation following recommended solvent systems and storage conditions.
  • Positive allosteric modulator of mGlu5 receptor, suitable for receptor pharmacology studies.
  • High purity (99.6%) for reliable experimental results.
  • Powder form for flexible dosing and formulation.
  • Soluble in DMSO at 50 mg/mL; recommended vehicle progression for in vivo administration.
  • Stable under recommended storage conditions for extended shelf life in powder form.