Learn More
Medchemexpress LLC N-(1,3-diphenylpyrazol-5-yl)-4-nitrobenzamide | 890764-36-0 | MFCD12546140 | 99.6% | 384.39 g·mol⁻¹ | C22H16N4O3 | 100 MG
SDP
Catalog No. 5000241393
Encompass_Preferred
VU-29 is a small-molecule positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5), used in pharmacology and neuroscience research to potentiate receptor responses. It is characterized by low-nanomolar activity and high reported purity, and is provided as a solid appropriate for in vitro receptor modulation studies.
  • Positive allosteric modulation of mGlu5 receptor (EC50 ≈ 9 nM).
  • Selective activity with reported Ki ≈ 244 nM for rmGluR5.
  • High reported purity suitable for research applications.
  • Available in multiple package sizes for experimental flexibility.
  • Small molecule with formula C22H16N4O3 and molecular weight 384.39 g·mol⁻¹.
Catalog No. 50-002-41393 Supplier Medchemexpress LLC Supplier No. HY107508100MG
VU-29 is a small-molecule positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5), used in pharmacology and neuroscience research to potentiate receptor responses. It is characterized by low-nanomolar activity and high reported purity, and is provided as a solid appropriate for in vitro receptor modulation studies.
  • Positive allosteric modulation of mGlu5 receptor (EC50 ≈ 9 nM).
  • Selective activity with reported Ki ≈ 244 nM for rmGluR5.
  • High reported purity suitable for research applications.
  • Available in multiple package sizes for experimental flexibility.
  • Small molecule with formula C22H16N4O3 and molecular weight 384.39 g·mol⁻¹.