Learn More
Medchemexpress LLC N-(1,3-diphenylpyrazol-5-yl)-4-nitrobenzamide | 890764-36-0 | MFCD12546140 | 99.6% | 384.39 g/mol | C22H16N4O3 | 5 MG
SDP
Catalog No. 5000240659
Encompass_Preferred
VU-29 is a research small molecule that acts as a positive allosteric modulator of the metabotropic glutamate 5 (mGlu5) receptor. It potentiates mGlu5 signaling with high potency and selectivity, and is supplied for laboratory research in solid or solution formats.
  • Positive allosteric modulator of mGlu5 with high potency (EC50 ≈ 9 nM).
  • Selective for mGlu5 versus other metabotropic glutamate receptors.
  • Reported high purity suitable for biochemical and cellular assays.
  • Available as small solid quantities or as 10 mM solutions in DMSO for convenience.
  • Chemical formula C22H16N4O3; molecular weight 384.39 g/mol.
Catalog No. 50-002-40659 Supplier Medchemexpress LLC Supplier No. HY1075085MG
VU-29 is a research small molecule that acts as a positive allosteric modulator of the metabotropic glutamate 5 (mGlu5) receptor. It potentiates mGlu5 signaling with high potency and selectivity, and is supplied for laboratory research in solid or solution formats.
  • Positive allosteric modulator of mGlu5 with high potency (EC50 ≈ 9 nM).
  • Selective for mGlu5 versus other metabotropic glutamate receptors.
  • Reported high purity suitable for biochemical and cellular assays.
  • Available as small solid quantities or as 10 mM solutions in DMSO for convenience.
  • Chemical formula C22H16N4O3; molecular weight 384.39 g/mol.