Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[1-[5-(3-chlorophenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]piperidin-4-yl]acetamide | 2869148-13-8 | 98.5% | 369.85 g/mol | C19H20ClN5O | 5 MG
SDP

Catalog No. 5000262616
Encompass_Preferred

SBP-3264 is a research-grade chemical probe that selectively inhibits Hippo-pathway kinases STK3 and STK4 and is used to investigate their roles in acute myeloid leukemia (AML). Supplied as a high-purity solid, it is intended for in vitro biochemical and cellular studies to probe pathway activity and disease-relevant signaling.

  • Selective STK3 and STK4 inhibition with low-nanomolar potency.
  • High purity suitable for biochemical and cellular assays.
  • Solid form with off-white to light yellow appearance.
  • Storage stability under recommended conditions (4°C; in solvent -80°C up to 6 months).
  • Soluble in DMSO for assay preparation and dilutions.

Catalog No. 50-002-62616 Supplier Medchemexpress LLC Supplier No. HY1329695MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SBP-3264 is a research-grade chemical probe that selectively inhibits Hippo-pathway kinases STK3 and STK4 and is used to investigate their roles in acute myeloid leukemia (AML). Supplied as a high-purity solid, it is intended for in vitro biochemical and cellular studies to probe pathway activity and disease-relevant signaling.

  • Selective STK3 and STK4 inhibition with low-nanomolar potency.
  • High purity suitable for biochemical and cellular assays.
  • Solid form with off-white to light yellow appearance.
  • Storage stability under recommended conditions (4°C; in solvent -80°C up to 6 months).
  • Soluble in DMSO for assay preparation and dilutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.