Learn More
Medchemexpress LLC N'-(1,8-dimethylimidazo[1,2-a]quinoxalin-4-yl)ethane-1,2-diamine | 445430-58-0 | MFCD18089812 | 99.7% | 255.32 g·mol⁻¹ | C14H17N5 | 10 MG
SDP
Catalog No. 5000242738
Encompass_Preferred
A selective, cell-permeable small-molecule inhibitor of IκB kinase that binds an allosteric site and preferentially inhibits IKK-2 (IC50 = 0.3 μM) over IKK-1 (IC50 = 4 μM). Supplied as a high-purity white to off-white solid for research use only, with supplier storage guidance for powder and solutions.
  • Selective inhibition of IKK-2 over IKK-1 (IC50 values reported)
  • Binds an allosteric site on IKK, useful for mechanistic studies
  • High purity solid suitable for biochemical and cell-based assays
  • Available in small pack sizes for screening and dose-response work
  • Stable when stored according to supplier guidance (powder and solvent conditions)
Catalog No. 50-002-42738 Supplier Medchemexpress LLC Supplier No. HY1051910MG
A selective, cell-permeable small-molecule inhibitor of IκB kinase that binds an allosteric site and preferentially inhibits IKK-2 (IC50 = 0.3 μM) over IKK-1 (IC50 = 4 μM). Supplied as a high-purity white to off-white solid for research use only, with supplier storage guidance for powder and solutions.
  • Selective inhibition of IKK-2 over IKK-1 (IC50 values reported)
  • Binds an allosteric site on IKK, useful for mechanistic studies
  • High purity solid suitable for biochemical and cell-based assays
  • Available in small pack sizes for screening and dose-response work
  • Stable when stored according to supplier guidance (powder and solvent conditions)