Learn More
Medchemexpress LLC N-(1-adamantyl)quinoxaline-2-carboxamide | 226878-01-9 | MFCD05865239 | 99.9% | 307.39 g·mol⁻¹ | C19H21N3O | 5 MG
SDP
Catalog No. 5000264534
Encompass_Preferred
NPS 2390 is a small-molecule research compound described as a noncompetitive antagonist of metabotropic glutamate receptors mGluR1 and mGluR5 and as an inhibitor of the calcium-sensing receptor (CaSR). It is supplied for laboratory research use only and is accompanied by technical documentation including a datasheet, COA, and SDS.
  • Noncompetitive antagonist of mGluR1 and mGluR5.
  • Inhibitor of the calcium-sensing receptor (CaSR).
  • Molecular formula C19H21N3O; molecular weight 307.39 g·mol⁻¹.
  • High purity (~99.9%) suitable for biochemical and cellular assays.
  • Available as solid quantities and as a 10 mM solution in DMSO.
  • Supplied for research use only with supporting COA, SDS, and handling instructions.
Catalog No. 50-002-64534 Supplier Medchemexpress LLC Supplier No. HY110955MG
NPS 2390 is a small-molecule research compound described as a noncompetitive antagonist of metabotropic glutamate receptors mGluR1 and mGluR5 and as an inhibitor of the calcium-sensing receptor (CaSR). It is supplied for laboratory research use only and is accompanied by technical documentation including a datasheet, COA, and SDS.
  • Noncompetitive antagonist of mGluR1 and mGluR5.
  • Inhibitor of the calcium-sensing receptor (CaSR).
  • Molecular formula C19H21N3O; molecular weight 307.39 g·mol⁻¹.
  • High purity (~99.9%) suitable for biochemical and cellular assays.
  • Available as solid quantities and as a 10 mM solution in DMSO.
  • Supplied for research use only with supporting COA, SDS, and handling instructions.