Learn More
Medchemexpress LLC N-(1-benzyl-2,3-dihydro-1H-pyrrolo[2,3-b]quinolin-4-yl)-2-phenylacetamide | 365565-02-2 | 99.3% | 25 MG
SDP
Catalog No. 5000234673
Encompass_Preferred
PGP-4008 is a selective P-glycoprotein (Pgp) inhibitor used in research to study multidrug resistance and Pgp-mediated drug transport in cellular and animal models.
  • Selective Pgp inhibitor for multidrug resistance research.
  • Reported purity approximately 99.3% by primary supplier.
  • CAS number 365565-02-2; molecular formula C26H23N3O; molecular weight 393.5 g/mol.
  • Typically supplied as a 25 MG solid; soluble in DMSO.
  • For research use only; follow appropriate handling and storage.
Catalog No. 50-002-34673 Supplier Medchemexpress LLC Supplier No. HY11982325MG
PGP-4008 is a selective P-glycoprotein (Pgp) inhibitor used in research to study multidrug resistance and Pgp-mediated drug transport in cellular and animal models.
  • Selective Pgp inhibitor for multidrug resistance research.
  • Reported purity approximately 99.3% by primary supplier.
  • CAS number 365565-02-2; molecular formula C26H23N3O; molecular weight 393.5 g/mol.
  • Typically supplied as a 25 MG solid; soluble in DMSO.
  • For research use only; follow appropriate handling and storage.