Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(1-benzyl-2,3-dihydropyrrolo[2,3-b]quinolin-4-yl)-2-phenylacetamide | 365565-02-2 | 99.3% | 100 MG
SDP

Catalog No. 5000234681
Encompass_Preferred

PGP-4008 is a selective P-glycoprotein (Pgp) inhibitor intended for research use. It has been shown to inhibit tumor growth in murine models of Pgp-mediated multidrug resistance when co-administered with doxorubicin.

  • Selective P-glycoprotein inhibitor
  • CAS 365565-02-2
  • Purity 99.26%
  • Molecular formula C26H23N3O
  • Molecular weight 393.48 g/mol
  • Available quantity 100 MG
  • Provided as a powder; soluble in DMSO

Catalog No. 50-002-34681 Supplier Medchemexpress LLC Supplier No. HY119823100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PGP-4008 is a selective P-glycoprotein (Pgp) inhibitor intended for research use. It has been shown to inhibit tumor growth in murine models of Pgp-mediated multidrug resistance when co-administered with doxorubicin.

  • Selective P-glycoprotein inhibitor
  • CAS 365565-02-2
  • Purity 99.26%
  • Molecular formula C26H23N3O
  • Molecular weight 393.48 g/mol
  • Available quantity 100 MG
  • Provided as a powder; soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.