Learn More
Cayman Chemical N- 1-Carboxyethyl-L-lysIn1mg
SDP
Catalog No. 502932558
Encompass_Preferred
An advanced glycation end product; protein bound, but not free CEL, binds to RAGE; elevated in the lens of diabetic patients with cataract; decreases glutamate uptake and secretion of S100B in rat hippocampal slices in a RAGE-independent manner at 1 mM
Catalog No. 50-293-2558 Supplier Cayman Chemical Supplier No. 253331 MG
An advanced glycation end product; protein bound, but not free CEL, binds to RAGE; elevated in the lens of diabetic patients with cataract; decreases glutamate uptake and secretion of S100B in rat hippocampal slices in a RAGE-independent manner at 1 mM