Learn More
Medchemexpress LLC N-[[1-(dimethylamino)cyclopentyl]methyl]-3,5-dimethoxy-4-(phenylmethoxy)benzamide hydrochloride | 495076-64-7 | 99.9% | 448.98 g/mol | C24H33ClN2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1075275MG
Encompass_Preferred
Org 25543 hydrochloride is the hydrochloride salt of a selective and irreversible glycine transporter 2 (GlyT2) inhibitor (IC50 16 nM) used as a research reagent to study glycine transport and analgesic mechanisms in preclinical models. It is supplied as a solid, high-purity material and should be stored sealed at -20°C to preserve stability.
  • Selective GlyT2 inhibition (IC50 16 nM).
  • Demonstrated analgesic activity in rodent neuropathic pain models.
  • High purity (>99.9%).
  • Solid form suitable for formulation and dosing in preclinical work.
  • Recommended storage at -20°C, sealed and protected from moisture.
  • Available in small research quantities for screening and pharmacology studies.
Catalog No. 50-002-40661