Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(1-methyl-1H-indol-5-yl)-N'-(3-methyl-5-isothiazolyl)urea | 152239-46-8 | MFCD00923644 | 99.9% | 286.35 g/mol | C14H14N4OS | 5 MG
SDP

Catalog No. 5000246766
Encompass_Preferred

SB 204741 is a selective, high-affinity antagonist of the serotonin 5-HT2B receptor supplied as an analytical standard for research applications. It has a reported pKi of 7.1, molecular formula C14H14N4OS, molecular weight 286.35 g/mol, and CAS 152239-46-8.

  • Selective 5-HT2B receptor antagonist (pKi = 7.1).
  • Provided as an analytical standard for research use.
  • Molecular formula C14H14N4OS; molecular weight 286.35 g/mol.
  • Soluble in DMSO: 100 mg/mL (may require ultrasonic assistance).
  • Suitable for pharmacology and receptor binding assays.
  • Store protected from light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-46766 Supplier Medchemexpress LLC Supplier No. HY1031535MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SB 204741 is a selective, high-affinity antagonist of the serotonin 5-HT2B receptor supplied as an analytical standard for research applications. It has a reported pKi of 7.1, molecular formula C14H14N4OS, molecular weight 286.35 g/mol, and CAS 152239-46-8.

  • Selective 5-HT2B receptor antagonist (pKi = 7.1).
  • Provided as an analytical standard for research use.
  • Molecular formula C14H14N4OS; molecular weight 286.35 g/mol.
  • Soluble in DMSO: 100 mg/mL (may require ultrasonic assistance).
  • Suitable for pharmacology and receptor binding assays.
  • Store protected from light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.