Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Methyl-3-phenylpropyl)valeramide, TCI America™
SDP

Catalog No. M097210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M097210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M097210G Supplier TCI America Supplier No. M097210G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 143086-32-2
Molecular Formula C15H23NO
Molecular Weight (g/mol) 233.355
MDL Number MFCD00144821
InChI Key PLWKOZGQPUWQNJ-UHFFFAOYSA-N
Synonym 1-Phenyl-3-valeramidobutane
PubChem CID 4657091
IUPAC Name N-(4-phenylbutan-2-yl)pentanamide
SMILES CCCCC(=O)NC(C)CCC1=CC=CC=C1

Specifications

Quantity 10 g
Formula Weight 233.36
Physical Form Liquid
Chemical Name or Material N-(1-Methyl-3-phenylpropyl)valeramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.