Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Naphthyl)-Ethylenediamine Dihydrochloride, 0.1%, for Nitrogen (Nitrite), LabChem™

Catalog No. LC175507 Shop All LabChem Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC175507 125 mL
LC175501 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC175507 Supplier LabChem, Inc. Supplier No. LC175507
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1465-25-4 , 7732-18-5
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.174
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
Synonym n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
PubChem CID 15106
ChEBI CHEBI:53452
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl

Specifications

Color Colorless or Tan
CAS Max % 99.9, 0.1
Linear Formula C10H7NHCH2CH2NH2·2HCl
Solubility Information Soluble in water
Formula Weight 259.18
Identification Passes Test
Packaging White Poly
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Hydrogen chloride
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material N-(1-Naphthyl)-Ethylenediamine Dihydrochloride, 0.1%, for Nitrogen (Nitrite)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Refrigerate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.