Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(1-Naphthyl)ethylenediamine dihydrochloride

Catalog No. N912525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N912525G 25 g
N912510G 10 g
N9125100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N912525G Supplier Sigma Aldrich Supplier No. N912525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-(1-Naphthyl)ethylenediamine dihydrochloride

Specifications

Melting Point 194°C to 198°C (dec.) (lit.)
Molecular Formula C12H14N2 -+ 2 HCl
Linear Formula C10H7NHCH2CH2NH2 -+ 2 HCl
MDL Number MFCD00012556
Quantity 25 g
Synonym 2-(1-Naphthylamino)ethylamine dihydrochloride
Molecular Weight (g/mol) 259.17
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.