Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Naphthyl)ethylenediamine Dihydrochloride, 95%, Spectrum™ Chemical
SDP

Catalog No. 18604709 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-709 10 kg Amber Glass Bottle
18-604-707 100 g Amber Glass Bottle
18-604-708 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-604-709 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N103110KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

N1031, 1465-25-4, C12H14N2·2HCl

Spectrum™ Chemical N-(1-Naphthyl)ethylenediamine Dihydrochloride

Chemical Identifiers

CAS 1465-25-4
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.17
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
IUPAC Name dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 10 kg
Percent Purity 95%
Grade Reagent
Physical Form Crystals or Flakes
Chemical Name or Material N-(1-Naphthyl)ethylenediamine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.