Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Naphthyl)ethylenediamine dihydrochloride, 96%

Catalog No. p-7023522
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1716409 10 g
AAA1716418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1716409 Supplier Thermo Scientific Chemicals Supplier No. A1716409
Add to Cart

CAS: 1465-25-4 | C12H16Cl2N2 | 259.174 g/mol

N-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.

Solubility
Soluble in hot water, alcohol, dilute hydrochloric acid and acetone.

Notes
Hygroscopic. Light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, halogen acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1465-25-4
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.174
MDL Number MFCD00012556
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
Synonym n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
PubChem CID 15106
ChEBI CHEBI:53452
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl

Specifications

Melting Point 191°C to 199°C
Quantity 10 g
Beilstein 3707471
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,6409
Solubility Information Soluble in hot water,alcohol,dilute hydrochloric acid and acetone.
Formula Weight 259.18
Percent Purity 96%
Chemical Name or Material N-(1-Naphthyl)ethylenediamine dihydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 215-981-2
RTECSNumber KV5330000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.