Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Naphthyl)phthalamic Acid 99.0+%, TCI America™
SDP

Catalog No. N006710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N006710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N006710G Supplier TCI America Supplier No. N006710G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 132-66-1
Molecular Formula C18H13NO3
Molecular Weight (g/mol) 291.306
MDL Number MFCD00037725
InChI Key JXTHEWSKYLZVJC-UHFFFAOYSA-N
Synonym naptalam, grelutin, alanap, alanape, analape, dyanap, peach-thin, n-1-naphthylphthalamic acid, mor-cran, alanap 1
PubChem CID 8594
ChEBI CHEBI:60833
IUPAC Name 2-(naphthalen-1-ylcarbamoyl)benzoic acid
SMILES C1=CC=C2C(=C1)C=CC=C2NC(=O)C3=CC=CC=C3C(=O)O

Specifications

Melting Point 187°C
Quantity 10 g
Formula Weight 291.31
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(1-Naphthyl)phthalamic Acid
RTECSNumber TH7350000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.