Learn More
Medchemexpress LLC N-(1-phenylethyl)-2-(piperazin-1-yl)quinazolin-4-amine | 662164-09-2 | MFCD04077665 | 99.5% | 333.43 | C20H23N5 | 10MM 1ML
SDP
Catalog No. 5000237227
Encompass_Preferred
D3-βArr is a small-molecule positive allosteric modulator of the thyrotropin receptor intended for research use. It is supplied as a solid and as a 10 mM solution in DMSO, and is used in receptor pharmacology and functional assays to study β-arrestin translocation and TSHR signaling.
  • Positive allosteric modulator of the thyrotropin receptor (TSHR).
  • Supports beta-arrestin translocation and TSHR functional assays.
  • High purity (reported ~99.5%).
  • Available as solid (multiple mg sizes) and 10 mM solution in DMSO.
  • Molecular formula C20H23N5, molecular weight 333.43.
Catalog No. 50-002-37227 Supplier Medchemexpress LLC Supplier No. HY12486710MM 1ML
D3-βArr is a small-molecule positive allosteric modulator of the thyrotropin receptor intended for research use. It is supplied as a solid and as a 10 mM solution in DMSO, and is used in receptor pharmacology and functional assays to study β-arrestin translocation and TSHR signaling.
  • Positive allosteric modulator of the thyrotropin receptor (TSHR).
  • Supports beta-arrestin translocation and TSHR functional assays.
  • High purity (reported ~99.5%).
  • Available as solid (multiple mg sizes) and 10 mM solution in DMSO.
  • Molecular formula C20H23N5, molecular weight 333.43.