Learn More
Medchemexpress LLC N-(1-phenylethyl)-2-(piperazin-1-yl)quinazolin-4-amine | 662164-09-2 | MFCD04077665 | 99.5% | 333.43 g/mol | C20H23N5 | 50MG
SDP
Catalog No. 5000237806
Encompass_Preferred
D3-βArr (N-(1-phenylethyl)-2-(piperazin-1-yl)quinazolin-4-amine; CAS 662164-09-2) is a small-molecule thyroid-stimulating-hormone (TSH) receptor modulator supplied as a high-purity solid for research use. It is provided in laboratory-scale quantities suitable for biochemical and cellular assays investigating TSH receptor pharmacology.
  • High reported purity (99.5%).
  • Solid form, white to off-white, suitable for standard storage.
  • Molecular weight 333.43 g/mol and formula C20H23N5.
  • Supplied in a 50 mg laboratory-scale quantity.
  • Intended for research use only; not for human or clinical use.
  • Suitable for biochemical and cellular assays targeting TSH receptor activity.
Catalog No. 50-002-37806 Supplier Medchemexpress LLC Supplier No. HY12486750MG
D3-βArr (N-(1-phenylethyl)-2-(piperazin-1-yl)quinazolin-4-amine; CAS 662164-09-2) is a small-molecule thyroid-stimulating-hormone (TSH) receptor modulator supplied as a high-purity solid for research use. It is provided in laboratory-scale quantities suitable for biochemical and cellular assays investigating TSH receptor pharmacology.
  • High reported purity (99.5%).
  • Solid form, white to off-white, suitable for standard storage.
  • Molecular weight 333.43 g/mol and formula C20H23N5.
  • Supplied in a 50 mg laboratory-scale quantity.
  • Intended for research use only; not for human or clinical use.
  • Suitable for biochemical and cellular assays targeting TSH receptor activity.