Learn More
Medchemexpress LLC N-(1-phenylethyl)-2-pyrrolidin-1-ylquinazolin-4-amine | 662163-81-7 | MFCD04077630 | 99.9% | 318.42 g·mol^-1 | C20H22N4 | 200 MG
SDP
Catalog No. 5000213036
Encompass_Preferred
Importazole is a small-molecule inhibitor of the nuclear transport receptor importin-β used in laboratory research to selectively block importin-β-mediated nuclear import. It is supplied as a white to off-white solid with documented solubility and storage recommendations for reliable use in cell biology studies. For research use only.
  • Selective inhibitor of importin-β.
  • High purity, typically about 99.9%.
  • White to off-white solid form.
  • Soluble in DMSO; in vitro and in vivo formulation protocols available.
  • Stable under recommended storage conditions for powder and solutions.
  • Intended for research use only; not for therapeutic use.
Catalog No. 50-002-13036 Supplier Medchemexpress LLC Supplier No. HY101091200MG
Importazole is a small-molecule inhibitor of the nuclear transport receptor importin-β used in laboratory research to selectively block importin-β-mediated nuclear import. It is supplied as a white to off-white solid with documented solubility and storage recommendations for reliable use in cell biology studies. For research use only.
  • Selective inhibitor of importin-β.
  • High purity, typically about 99.9%.
  • White to off-white solid form.
  • Soluble in DMSO; in vitro and in vivo formulation protocols available.
  • Stable under recommended storage conditions for powder and solutions.
  • Intended for research use only; not for therapeutic use.