Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(1-Pyrenyl)maleimide

Catalog No. P79081G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P79081G 1 g
P7908500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P79081G Supplier Sigma Aldrich Supplier No. P79081G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-(1-Pyrenyl)maleimide

Specifications

CAS 42189-56-0
Melting Point 235°C to 237°C (lit.)
Molecular Formula C20H11NO2
Linear Formula C20H11NO2
MDL Number MFCD00049301
Quantity 1 g
Synonym 1-(1-Pyrenyl)-1 H-pyrrole-2,5-dione
Molecular Weight (g/mol) 297.31
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.