Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Pyrenyl)maleimide 97.0+%, TCI America™
SDP

Catalog No. P1214250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1214250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1214250MG Supplier TCI America Supplier No. P1214250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 42189-56-0
Molecular Formula C20H11NO2
Molecular Weight (g/mol) 297.31
MDL Number MFCD00049301
InChI Key NPTUGEKDRBZJRE-UHFFFAOYSA-N
Synonym n-1-pyrenyl maleimide, n-1-pyrene maleimide, 1-pyren-1-yl-1h-pyrrole-2,5-dione, n-3-pyrenyl maleimide, 1-pyren-1-yl pyrrole-2,5-dione, n-1-pyrenyl-maleimide, acmc-209jo6, n-3-pyrene maleeimide, 1h-pyrrole-2,5-dione,1-1-pyrenyl
PubChem CID 626783
IUPAC Name 1-(pyren-4-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
SMILES O=C1C=CC(=O)N1C1=CC2=CC=CC3=CC=C4C=CC=C1C4=C23

Specifications

Quantity 250 mg
Formula Weight 297.31
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(1-Pyrenyl)maleimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.