Learn More
Biotium N-(1, 10-phenanthrolin-5-yl)iodoacetamide
Catalog No. 501964841
Encompass_Preferred
N-(1,10-Phenanthrolin-5-yl)iodoacetamide has been used to label thiols to confer the metal-binding properties of the phenanthroline chelator. For example, completing of the chelator with copper can be used to selectively cleave RNA or DNA in the presence of hydrogen peroxide. Properties: Off-white solid soluble in DMF; Store at -20°C; 14H10IN3O; MW: 363.15.
Catalog No. 50-196-4841 Supplier Biotium Supplier No. 92015
N-(1,10-Phenanthrolin-5-yl)iodoacetamide has been used to label thiols to confer the metal-binding properties of the phenanthroline chelator. For example, completing of the chelator with copper can be used to selectively cleave RNA or DNA in the presence of hydrogen peroxide. Properties: Off-white solid soluble in DMF; Store at -20°C; 14H10IN3O; MW: 363.15.