Learn More
Medchemexpress LLC N-[12-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]dodecanoyl]-sphinganine | 474943-05-0 | MFCD02066201 | >99.0% | C36H63N5O6 | 10MG
SDP
Catalog No. 5000260220
Encompass_Preferred
C12-NBD sphinganine is an NBD-labeled sphingolipid used as a fluorescent substrate for biochemical assays. It is cleaved by ceramidases, enabling quantitative measurement of alkaline and neutral ceramidase activity in biological samples by fluorescence.
  • Fluorescent NBD label enables sensitive fluorescence-based detection.
  • Suitable for measurement of alkaline and neutral ceramidase activity.
  • High purity (>99.0%).
  • Chemical formula C36H63N5O6; molecular weight 661.92 g·mol⁻¹.
  • Supplied as a powder; store at -20°C to maintain stability.
Catalog No. 50-002-60220 Supplier Medchemexpress LLC Supplier No. HY14157710MG
C12-NBD sphinganine is an NBD-labeled sphingolipid used as a fluorescent substrate for biochemical assays. It is cleaved by ceramidases, enabling quantitative measurement of alkaline and neutral ceramidase activity in biological samples by fluorescence.
  • Fluorescent NBD label enables sensitive fluorescence-based detection.
  • Suitable for measurement of alkaline and neutral ceramidase activity.
  • High purity (>99.0%).
  • Chemical formula C36H63N5O6; molecular weight 661.92 g·mol⁻¹.
  • Supplied as a powder; store at -20°C to maintain stability.