Learn More
Cayman Chemical Nε-(1-Carboxymethyl)-L-lysine, Purity: ≥95%, CAS: 5746-04-3, 10 mg
SDP
Catalog No. 502971947
Encompass_Preferred
An AGE, produced by the oxidative modification of glycated proteins during oxidative stress; increases with aging and during pathologies marked by oxidative stress; activates RAGE, triggering signaling through MAPKs and NF-κB
Catalog No. 50-297-1947 Supplier Cayman Chemical Supplier No. 1648310 MG
An AGE, produced by the oxidative modification of glycated proteins during oxidative stress; increases with aging and during pathologies marked by oxidative stress; activates RAGE, triggering signaling through MAPKs and NF-κB