Learn More
Medchemexpress LLC N-(1H-benzimidazol-2-yl)-3-(1H-pyrrol-1-yl)benzamide | 1190277-80-5 | MFCD13758448 | 98.1% | C18H14N4O | 10MG
SDP
Catalog No. 5000259397
Encompass_Preferred
MKI-1 is a small-molecule research inhibitor of MASTL (microtubule-associated serine/threonine kinase-like) that inhibits MASTL with an IC50 of 9.9 μM. It has demonstrated antitumor and radiosensitizing activity in breast cancer models through activation of PP2A and reduces c-Myc stability in cellular assays. Supplied as an off-white to light yellow solid with reported purity of ~98% and high DMSO solubility.
  • Inhibits MASTL with IC50 of 9.9 μM.
  • Demonstrates antitumor and radiosensitizer activity via PP2A activation.
  • Active in breast cancer cell assays at low micromolar concentrations.
  • Reduces c-Myc phosphorylation and stability in cells.
  • Off-white to light yellow solid with high reported purity (~98%).
  • Soluble in DMSO at high concentration (about 250 mg/mL).
  • Intended for in vitro research use only.
Catalog No. 50-002-59397 Supplier Medchemexpress LLC Supplier No. HY13755210MG
MKI-1 is a small-molecule research inhibitor of MASTL (microtubule-associated serine/threonine kinase-like) that inhibits MASTL with an IC50 of 9.9 μM. It has demonstrated antitumor and radiosensitizing activity in breast cancer models through activation of PP2A and reduces c-Myc stability in cellular assays. Supplied as an off-white to light yellow solid with reported purity of ~98% and high DMSO solubility.
  • Inhibits MASTL with IC50 of 9.9 μM.
  • Demonstrates antitumor and radiosensitizer activity via PP2A activation.
  • Active in breast cancer cell assays at low micromolar concentrations.
  • Reduces c-Myc phosphorylation and stability in cells.
  • Off-white to light yellow solid with high reported purity (~98%).
  • Soluble in DMSO at high concentration (about 250 mg/mL).
  • Intended for in vitro research use only.