Learn More
Medchemexpress LLC N-((1H-indol-3-yl)acetyl)-L-tryptophan | 57105-53-0 | MFCD18641983 | >98.0% | C21H19N3O3 | 10MG
SDP
Catalog No. 5000261618
Encompass_Preferred
(S)-N-(1H-indole-3-acetyl)tryptophan is the S-stereoisomer of N-(1H-indole-3-acetyl)tryptophan, a tryptophan-derived small molecule (CAS 57105-53-0) with molecular formula C21H19N3O3 and molar mass 361.4 g/mol. It is provided for research use and has been reported to weakly inhibit β-D-glucosidase.
  • Stereospecific small molecule with defined (S)-configuration.
  • Molecular formula C21H19N3O3, molar mass 361.4 g/mol.
  • Listed with CAS number 57105-53-0 and standard chemical identifiers.
  • Reported weak inhibition of β-D-glucosidase in literature.
  • Intended for research use; confirm purity and storage from supplier datasheet.
Catalog No. 50-002-61618 Supplier Medchemexpress LLC Supplier No. HY13881510MG
(S)-N-(1H-indole-3-acetyl)tryptophan is the S-stereoisomer of N-(1H-indole-3-acetyl)tryptophan, a tryptophan-derived small molecule (CAS 57105-53-0) with molecular formula C21H19N3O3 and molar mass 361.4 g/mol. It is provided for research use and has been reported to weakly inhibit β-D-glucosidase.
  • Stereospecific small molecule with defined (S)-configuration.
  • Molecular formula C21H19N3O3, molar mass 361.4 g/mol.
  • Listed with CAS number 57105-53-0 and standard chemical identifiers.
  • Reported weak inhibition of β-D-glucosidase in literature.
  • Intended for research use; confirm purity and storage from supplier datasheet.