Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[(1R)-1-(3-cyclopentyloxyphenyl)ethyl]-3-[(2,4-dioxopyrimidin-1-yl)methoxy]propane-1-sulfonamide | 1198221-21-4 | 99.9% | 451.54 g/mol | C21H29N3O6S | 10MG
SDP

Catalog No. 5000236809
Encompass_Preferred

TAS-114 is a small-molecule, orally active dual inhibitor of dUTPase and dihydropyrimidine dehydrogenase (DPD) used in preclinical and clinical research to enhance the efficacy of fluoropyrimidine-based chemotherapy and to probe nucleotide metabolism in cancer models.

  • Dual inhibition of dUTPase and DPD for combination chemotherapy studies.
  • Orally active small molecule suitable for in vitro and in vivo research.
  • High reported purity (99.89%).
  • Molecular formula C21H29N3O6S; MW 451.54 g/mol.
  • Recommended storage: 4°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Analytical data available: COA, SDS, data sheet, and spectral data.

Catalog No. 50-002-36809 Supplier Medchemexpress LLC Supplier No. HY12406210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TAS-114 is a small-molecule, orally active dual inhibitor of dUTPase and dihydropyrimidine dehydrogenase (DPD) used in preclinical and clinical research to enhance the efficacy of fluoropyrimidine-based chemotherapy and to probe nucleotide metabolism in cancer models.

  • Dual inhibition of dUTPase and DPD for combination chemotherapy studies.
  • Orally active small molecule suitable for in vitro and in vivo research.
  • High reported purity (99.89%).
  • Molecular formula C21H29N3O6S; MW 451.54 g/mol.
  • Recommended storage: 4°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Analytical data available: COA, SDS, data sheet, and spectral data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.