Learn More
Medchemexpress LLC N-[(1R)-1-(3-cyclopentyloxyphenyl)ethyl]-3-[(2,4-dioxopyrimidin-1-yl)methoxy]propane-1-sulfonamide | 1198221-21-4 | 99.9% | 451.54 g/mol | C21H29N3O6S | 10MM 1ML
SDP
Catalog No. 5000238384
Encompass_Preferred
TAS-114 is an orally active dual inhibitor of dUTPase and dihydropyrimidine dehydrogenase (DPD) used in research to potentiate the effects of fluoropyrimidine chemotherapy. Supplied as a 10 mM solution in DMSO (1 mL), the compound has CAS 1198221-21-4, molecular formula C21H29N3O6S, molecular weight 451.54 g/mol, and high purity suitable for biochemical and cell-based studies.
  • Supplied as a 10 mM solution in DMSO for ready-to-use dosing.
  • Dual inhibition of dUTPase and DPD to potentiate fluoropyrimidine activity.
  • High purity (99.9%) suitable for in vitro experiments.
  • Includes molecular formula and molecular weight for experimental reference.
  • Suitable for biochemical assays and cellular studies of fluoropyrimidine metabolism.
Catalog No. 50-002-38384 Supplier Medchemexpress LLC Supplier No. HY12406210MM 1ML
TAS-114 is an orally active dual inhibitor of dUTPase and dihydropyrimidine dehydrogenase (DPD) used in research to potentiate the effects of fluoropyrimidine chemotherapy. Supplied as a 10 mM solution in DMSO (1 mL), the compound has CAS 1198221-21-4, molecular formula C21H29N3O6S, molecular weight 451.54 g/mol, and high purity suitable for biochemical and cell-based studies.
  • Supplied as a 10 mM solution in DMSO for ready-to-use dosing.
  • Dual inhibition of dUTPase and DPD to potentiate fluoropyrimidine activity.
  • High purity (99.9%) suitable for in vitro experiments.
  • Includes molecular formula and molecular weight for experimental reference.
  • Suitable for biochemical assays and cellular studies of fluoropyrimidine metabolism.