Learn More
Medchemexpress LLC N-[(1R)-1-(4-Fluorophenyl)ethyl]-3-[2-(isoquinolin-3-yl)-1H-imidazol-4-yl]benzamide | 950225-08-8 | 99.9% | 436.48 g/mol | C27H21FN4O | 100MG
SDP
Catalog No. 5000239139
Encompass_Preferred
TTP-8307 is a small-molecule inhibitor used in laboratory research that inhibits replication of certain rhino- and enteroviruses and shows activity against hepatitis C virus. It is supplied in multiple sizes and formats for research purposes only and is not intended for human use.
  • Inhibits replication of rhino- and enteroviruses and shows activity against hepatitis C virus.
  • Supplied for research use only and not for clinical or human use.
  • Available as solid quantities and as DMSO solutions for convenience in assays.
  • High purity supports reproducible biochemical and cellular studies.
  • Molecular formula C27H21FN4O and molecular weight 436.48 g/mol.
  • Multiple package sizes available, with larger quantities provided by quotation.
Catalog No. 50-002-39139 Supplier Medchemexpress LLC Supplier No. HY124806100MG
TTP-8307 is a small-molecule inhibitor used in laboratory research that inhibits replication of certain rhino- and enteroviruses and shows activity against hepatitis C virus. It is supplied in multiple sizes and formats for research purposes only and is not intended for human use.
  • Inhibits replication of rhino- and enteroviruses and shows activity against hepatitis C virus.
  • Supplied for research use only and not for clinical or human use.
  • Available as solid quantities and as DMSO solutions for convenience in assays.
  • High purity supports reproducible biochemical and cellular studies.
  • Molecular formula C27H21FN4O and molecular weight 436.48 g/mol.
  • Multiple package sizes available, with larger quantities provided by quotation.