Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-[(1R,2R)-2-(1-Piperidinyl)Cyclohexyl]-N'-[4-(Trifluoromethyl)Phenyl]Squaramide

Catalog No. 728357100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-835-7100MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 72-835-7100MG Supplier Sigma Aldrich Supplier No. 728357100MG
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

It can act as a catalyst for the asymmetric tandem aza-Michael addition-protonation between 1-nitro cyclohexene and 4-methoxyaniline to form the corresponding +a-arylamino-+a-nitro cyclohexane in a 35:65 trans/cis mixture.

Specifications

CAS 454482-11-2
Molecular Formula C40H81N3O20
Linear Formula C40H81N3O20
MDL Number MFCD13184941
Quantity 250 mg
Synonym Azido-PEG (n=19)
Molecular Weight (g/mol) 924.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.