Learn More
Medchemexpress LLC N-((1R,3S)-3-(4-acetylpiperazin-1-yl)cyclohexyl)-7-fluoro-4-methyl-1H-indole-2-carboxamide | 2411748-50-8 | 99.1% | C22H29FN4O2 | 10MG
SDP
Catalog No. 5000258545
Encompass_Preferred
EZM0414 is a selective, orally bioavailable small-molecule inhibitor of the histone methyltransferase SETD2, developed for preclinical research applications. It exhibits potent biochemical and cellular activity and is intended for laboratory research into SETD2-driven biology and oncology models.
  • Selective SETD2 inhibitor with biochemical IC50 ~18 nM.
  • Demonstrates cellular activity (IC50 ~34 nM).
  • Orally bioavailable in preclinical models.
  • High purity (≈99%) suitable for biochemical and cellular assays.
  • Applicable to research on hematologic and solid tumor models.
Catalog No. 50-002-58545 Supplier Medchemexpress LLC Supplier No. HY14485810MG
EZM0414 is a selective, orally bioavailable small-molecule inhibitor of the histone methyltransferase SETD2, developed for preclinical research applications. It exhibits potent biochemical and cellular activity and is intended for laboratory research into SETD2-driven biology and oncology models.
  • Selective SETD2 inhibitor with biochemical IC50 ~18 nM.
  • Demonstrates cellular activity (IC50 ~34 nM).
  • Orally bioavailable in preclinical models.
  • High purity (≈99%) suitable for biochemical and cellular assays.
  • Applicable to research on hematologic and solid tumor models.